Learn More
Trajan Scientific VICI Valco 10 Port Cheminert Switching UHP Valve, 15K PSI, 1.50mm (VAL.C72X-6690D-CTC)
Catalog No. 50635036
Encompass
VICI Valco VICI Valco Intruments PN- C72X-6690D-CTC, 15000 PSI, Valve Style- Cheminert, Ports- 10, Port Size- 1.50mm, 2 Positions Coated Stainless VICI Valco Replacement Components Stator PN- C72-6C90 LPP Stator PN- 100.LCC2VS10UHP VICI Valco Rotor PN- C72-66R0 LPP Rotor PN-100.LCC2VRUHP10 (NEW LPP PN: VAL.C72X-6690D-CTC). Thermo Part Number: 00950-01-00327. Made in the USA.
Catalog No. 50-635-036 Supplier Trajan Scientific Supplier No. PAL.VLV10UHPV
VICI Valco VICI Valco Intruments PN- C72X-6690D-CTC, 15000 PSI, Valve Style- Cheminert, Ports- 10, Port Size- 1.50mm, 2 Positions Coated Stainless VICI Valco Replacement Components Stator PN- C72-6C90 LPP Stator PN- 100.LCC2VS10UHP VICI Valco Rotor PN- C72-66R0 LPP Rotor PN-100.LCC2VRUHP10 (NEW LPP PN: VAL.C72X-6690D-CTC). Thermo Part Number: 00950-01-00327. Made in the USA.