Learn More
Grainger Schneider Electric Hydronic Globe Valve, 2-Way, Bronze, 40 GPM, 1 ea
Catalog No. 502761355
Encompass_Preferred
This 2-way hydronic globe valve is designed for reliable flow control in HVACR applications, such as hot or chilled water and steam systems. Featuring a 2 in. NPT connection and a flow coefficient (Cv) of 40, it is engineered for precision and efficiency in demanding environments. The valve utilizes a robust bronze body and stainless steel stem to ensure long-term durability and performance.
  • High rangeability provides fine, accurate control for efficient and responsive regulation.
  • Stem-up to close action with a soft seat design.
  • Suitable for water and steam media across a temperature range of -7°C to 138°C (20°F to 281°F).
  • Self-adjusting packing design with spring-loaded PTFE/EPDM.
  • Compatible with both pneumatic and electric actuators.
Catalog No. 50-276-1355 Supplier Grainger Supplier No. 161Z01
This 2-way hydronic globe valve is designed for reliable flow control in HVACR applications, such as hot or chilled water and steam systems. Featuring a 2 in. NPT connection and a flow coefficient (Cv) of 40, it is engineered for precision and efficiency in demanding environments. The valve utilizes a robust bronze body and stainless steel stem to ensure long-term durability and performance.
  • High rangeability provides fine, accurate control for efficient and responsive regulation.
  • Stem-up to close action with a soft seat design.
  • Suitable for water and steam media across a temperature range of -7°C to 138°C (20°F to 281°F).
  • Self-adjusting packing design with spring-loaded PTFE/EPDM.
  • Compatible with both pneumatic and electric actuators.