Learn More
Grainger Schneider Electric Hydronic Globe Valve, Bronze, 2 in, 40 Cv
Catalog No. 502760987
Encompass_Preferred
The VB-7283 series is a 2-way globe valve designed for the control of chilled water, hot water, or steam in HVAC applications. This threaded, bronze-bodied valve features a stem-up-to-close design with stainless steel trim, providing reliable performance in both two-position and proportional control systems. It is engineered for a temperature range of 20°F to 400°F (-7°C to 204°C) and supports a flow capacity of 40 Cv. The valve requires a compatible actuator and linkage, available separately, for operation.
  • Bronze body construction with stainless steel seat, plug, and stem
  • Self-adjusting TFE packing design
  • Two-way body pattern with threaded (NPT) connections
  • Designed for hot water, chilled water, and steam service
  • Suitable for two-position, floating, or proportional control applications
Catalog No. 50-276-0987 Supplier Grainger Supplier No. 161Z26
The VB-7283 series is a 2-way globe valve designed for the control of chilled water, hot water, or steam in HVAC applications. This threaded, bronze-bodied valve features a stem-up-to-close design with stainless steel trim, providing reliable performance in both two-position and proportional control systems. It is engineered for a temperature range of 20°F to 400°F (-7°C to 204°C) and supports a flow capacity of 40 Cv. The valve requires a compatible actuator and linkage, available separately, for operation.
  • Bronze body construction with stainless steel seat, plug, and stem
  • Self-adjusting TFE packing design
  • Two-way body pattern with threaded (NPT) connections
  • Designed for hot water, chilled water, and steam service
  • Suitable for two-position, floating, or proportional control applications