Learn More
Waters Corp Valve, 2-Way Solenoid, Replacement, UPLC, HPLC
Catalog No. 502392905 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
  • Replacement 2-way solenoid valve.
  • Utilized on 2414 and ACQUITY Refractive Index Detectors.
  • Compatible with UPLC and HPLC systems.
  • Replacement recommended as required.
Catalog No. 50-239-2905 Supplier Waters Corp Supplier No. 700002360
  • Replacement 2-way solenoid valve.
  • Utilized on 2414 and ACQUITY Refractive Index Detectors.
  • Compatible with UPLC and HPLC systems.
  • Replacement recommended as required.