Learn More
AVANTOR MASTERFLEX (VWR LLC) John Guest PPSV041212W Speedfit Ball Valve, straight, 3/8"" OD speedfit x 3/8"" OD speedfit
Catalog No. NC0895030
Encompass
Food-grade polypropylene housing is specifically made for use with potable water. Quickly disconnect and reconnect your water lines with these Speedfit valves.
Catalog No. NC0895030 Supplier AVANTOR MASTERFLEX (VWR LLC) Supplier No. MFLX0739106
Food-grade polypropylene housing is specifically made for use with potable water. Quickly disconnect and reconnect your water lines with these Speedfit valves.