Learn More
Chemglass Life Sciences Valve, Chem-Vac, Chem-Cap, Hi-Vac, Three-Way, 0-8mm Bore
SDP
Catalog No. CG95702
Encompass_Preferred
Valve, Chem-Vac, Chem-Cap, Hi-Vac, Three-Way, 0-8mm Bore. CHEM-VAC CHEM-CAP valve with three-way configuration for use in construction of manifolds, adapters, etc. See CG-961 for replacement CHEM-CAP plug and CG-906 for glass portion only.
Catalog No. CG95702 Supplier Chemglass Life Sciences Supplier No. CG95702
Valve, Chem-Vac, Chem-Cap, Hi-Vac, Three-Way, 0-8mm Bore. CHEM-VAC CHEM-CAP valve with three-way configuration for use in construction of manifolds, adapters, etc. See CG-961 for replacement CHEM-CAP plug and CG-906 for glass portion only.