Learn More
New Jersey Fluid Systems LLC VALVE
Catalog No. NC9382533
Encompass
Swagelok 42G series Ball Valve. Straight Pattern; 1/4 in. OD Tube Fitting ends; 2,500 psi max; 316 Stainless Steel
Catalog No. NC9382533 Supplier New Jersey Fluid Systems LLC Supplier No. SS42GS4
Swagelok 42G series Ball Valve. Straight Pattern; 1/4 in. OD Tube Fitting ends; 2,500 psi max; 316 Stainless Steel