Learn More
Trajan Scientific VICI Valco Cheminert 6 Port Injection Valve, 0.75mm Bore (VAL.C2V-3006D-CTC-K)
Catalog No. 50635040
Encompass
VICI Valco VICI Valco Intruments PN- C2V-3006D-CTC-K, 5000 PSI, Valve Style- Cheminert, Ports- 6, Port Size- 0.75 mm, 2 Positions Nitronic 60 (NEW LPP PN: VAL.C2V-3006D-CTC-K) VICI Valco Replacement Components Stator PN- C2V-3C06 LPP Stator PN- 100.LCC2VSPREP6 VICI Valco Rotor PN- C2-30R6 LPP Rotor PN-100.LCC2VRPREP6 Thermo Part Number: 00950-01-00320. Made in the USA.
Catalog No. 50-635-040 Supplier Trajan Scientific Supplier No. PAL.VLV6PREP
VICI Valco VICI Valco Intruments PN- C2V-3006D-CTC-K, 5000 PSI, Valve Style- Cheminert, Ports- 6, Port Size- 0.75 mm, 2 Positions Nitronic 60 (NEW LPP PN: VAL.C2V-3006D-CTC-K) VICI Valco Replacement Components Stator PN- C2V-3C06 LPP Stator PN- 100.LCC2VSPREP6 VICI Valco Rotor PN- C2-30R6 LPP Rotor PN-100.LCC2VRPREP6 Thermo Part Number: 00950-01-00320. Made in the USA.