Learn More
Trajan Scientific VICI Valco Valco 6 Port Injection Valve, Hastelloy (VAL.DC6WHC-K-LP)
Catalog No. 50635060
Encompass
VICI Valco Valve 6 Port 2 Position, Hastelloy C, 5K PSI, 0.40mm Ports.[VICI Valco PN: DC6WHC-K-LP]"VICI Valco Intruments Pressure 5000PSI, VICI Valco Style, 6 Ports, Port Size 0.40mm 2 Valve 6 Port 2 Position, 5K PSI, 0.40mm Ports. [VICI Valco PN: DC6W-K-LP] VICI Valco Part # DC6W-K-LP Replacement Rotor 100.LC26R " VICI Valco Part # DC6W-K-LP Replacement Rotor 100.LC26R " (NEW LPP PN: VAL.DC6WHC-K-LP). Made in the USA.
Catalog No. 50-635-060 Supplier Trajan Scientific Supplier No. 100.LC62H
VICI Valco Valve 6 Port 2 Position, Hastelloy C, 5K PSI, 0.40mm Ports.[VICI Valco PN: DC6WHC-K-LP]"VICI Valco Intruments Pressure 5000PSI, VICI Valco Style, 6 Ports, Port Size 0.40mm 2 Valve 6 Port 2 Position, 5K PSI, 0.40mm Ports. [VICI Valco PN: DC6W-K-LP] VICI Valco Part # DC6W-K-LP Replacement Rotor 100.LC26R " VICI Valco Part # DC6W-K-LP Replacement Rotor 100.LC26R " (NEW LPP PN: VAL.DC6WHC-K-LP). Made in the USA.