Learn More
Trajan Scientific VICI Valco Valco 6 Port Injection Valve 0.40 Stainless Steel (VAL.DC6W-K-LP)
Catalog No. 50635061
Encompass
VICI Valco VICI Valco Intruments PN- DC6W-K-LP, 5000 PSI, Valve Style- VICI Valco, Ports- 6, Port Size- 0.40mm, 2 Positions (NEW LPP PN: VAL.DC6W-K-LP)VICI Valco Replacement Components Stator PN- None LPP Stator PN- None VICI Valco Rotor PN- SSAC6W LPP Rotor PN-100.LC26R. Made in the USA.
Catalog No. 50-635-061 Supplier Trajan Scientific Supplier No. 100.LC62S
VICI Valco VICI Valco Intruments PN- DC6W-K-LP, 5000 PSI, Valve Style- VICI Valco, Ports- 6, Port Size- 0.40mm, 2 Positions (NEW LPP PN: VAL.DC6W-K-LP)VICI Valco Replacement Components Stator PN- None LPP Stator PN- None VICI Valco Rotor PN- SSAC6W LPP Rotor PN-100.LC26R. Made in the USA.