Learn More
Trajan Scientific VICI Valco Cheminert 6 Port Injection Valve, 0.40mm (VAL.C2V-2006D-CTC)
Catalog No. 50635037
Encompass
VICI Valco Valve 6 Port 2 Position, 5K PSI, 0.40mm Ports, Cheminert [VICI Valco PN: C2V-2006D-CTC] (NEW LPP PN: VAL.C2V-2006D-CTC). Thermo Part Number: 00950-01-00319. Made in the USA.
Catalog No. 50-635-037 Supplier Trajan Scientific Supplier No. PAL.VLV6
VICI Valco Valve 6 Port 2 Position, 5K PSI, 0.40mm Ports, Cheminert [VICI Valco PN: C2V-2006D-CTC] (NEW LPP PN: VAL.C2V-2006D-CTC). Thermo Part Number: 00950-01-00319. Made in the USA.