Learn More
AVANTOR MASTERFLEX (VWR LLC) Sanitary Ball Valve, 316 SS and PTFE, 3/4""
Catalog No. NC9028969
Encompass
Ball valves feature a unique design that allows for easy in-line servicing. Valve seats are made of fully encapsulated PTFE. Each valve includes a locking handle.
Catalog No. NC9028969 Supplier AVANTOR MASTERFLEX (VWR LLC) Supplier No. MFLX3053000
Ball valves feature a unique design that allows for easy in-line servicing. Valve seats are made of fully encapsulated PTFE. Each valve includes a locking handle.