Learn More
Grainger Murdock Valve Cartridge Assembly, For Drinking Fountains, 1 ea
Catalog No. 502777323
Encompass_Preferred
This valve cartridge assembly is a replacement component designed for use with drinking fountains. It helps regulate water flow and ensures proper functionality within compatible plumbing systems.
  • Includes 0.5 GPM flow regulator cartridge, 1/4 in. OD insert assembly, brass sleeve, jam nut, regulator valve body, and retaining nut
  • Compatible with specific drinking fountains
  • Complies with Federal Public Law 111-380 (no lead)
  • Tested in compliance with ARI 1010
  • Overall dimensions: 2-3/16 in. width by 2-1/4 in. height by 1-5/8 in. depth
Catalog No. 50-277-7323 Supplier Grainger Supplier No. 189L43
This valve cartridge assembly is a replacement component designed for use with drinking fountains. It helps regulate water flow and ensures proper functionality within compatible plumbing systems.
  • Includes 0.5 GPM flow regulator cartridge, 1/4 in. OD insert assembly, brass sleeve, jam nut, regulator valve body, and retaining nut
  • Compatible with specific drinking fountains
  • Complies with Federal Public Law 111-380 (no lead)
  • Tested in compliance with ARI 1010
  • Overall dimensions: 2-3/16 in. width by 2-1/4 in. height by 1-5/8 in. depth