Learn More
Med Vet International Centesis Valve w/ Drain , J1339G, Each
Catalog No. 5000495619
Encompass_Preferred
Bivalve allows syringe to aspirate fluids i.e. chest drainage and then expel those fluids via another port so continually active centesis can be achieved without disconnecting. Suggest use J0282d1 Barbed Connector for chest drainage or feeding tube connection. Centesis Drainage Kit spin luerlock attaches directly to bivalv
Catalog No. 50-004-95619 Supplier Med Vet International Supplier No. JOR1339G
Bivalve allows syringe to aspirate fluids i.e. chest drainage and then expel those fluids via another port so continually active centesis can be achieved without disconnecting. Suggest use J0282d1 Barbed Connector for chest drainage or feeding tube connection. Centesis Drainage Kit spin luerlock attaches directly to bivalv