Learn More
Chemglass Life Sciences Valve, Chem-Vac, Glass Plug, Hi-Vac, Low Hold-Up, 0-4mm Bore, Kalrez O-Ring
SDP
Catalog No. 501531875
Encompass_Preferred
Valve, Chem-Vac, Glass Plug, Hi-Vac, Low Hold-Up, 0-4mm Bore, Kalrez O-Ring. Similar to CG-975 CHEM-VAC valve but with the arms bent to minimize hold up of fluids. Valve has a glass plug with three o-rings, the tip o-ring making the primary seal on the precision formed tapered seat. Available with a choice of Viton or Kalrez o-rings. See CG-983 for replacement plug and CG-585 for o-ring replacement kit. If individual o-ring is required see CG-305 for Viton or CG-308 for Kalrez. For replacement glass portion of valve only see CG-905.
Catalog No. 50-153-1875 Supplier Chemglass Life Sciences Supplier No. CG97730
Valve, Chem-Vac, Glass Plug, Hi-Vac, Low Hold-Up, 0-4mm Bore, Kalrez O-Ring. Similar to CG-975 CHEM-VAC valve but with the arms bent to minimize hold up of fluids. Valve has a glass plug with three o-rings, the tip o-ring making the primary seal on the precision formed tapered seat. Available with a choice of Viton or Kalrez o-rings. See CG-983 for replacement plug and CG-585 for o-ring replacement kit. If individual o-ring is required see CG-305 for Viton or CG-308 for Kalrez. For replacement glass portion of valve only see CG-905.