Learn More
Chemglass Life Sciences Valve, Chem-Vac, Chem-Cap, Hi-Vac, Metering, Low Hold-Up, 0-4mm Bore
SDP
Catalog No. 501531872
Encompass_Preferred
Valve, Chem-Vac, Chem-Cap, Hi-Vac, Metering, Low Hold-Up, 0-4mm Bore. CHEM-VAC CHEM-CAP metering valve has an extended tip on the plug to allow semi-needle valve control of gases and liquids. Similar to CG-965 CHEM-VAC valve but with the arms bent to minimize hold-up of fluids. See CG-971 for replacement CHEM-CAP metering plug and CG-905 for glass portion only.
Catalog No. 50-153-1872 Supplier Chemglass Life Sciences Supplier No. CG96701
Valve, Chem-Vac, Chem-Cap, Hi-Vac, Metering, Low Hold-Up, 0-4mm Bore. CHEM-VAC CHEM-CAP metering valve has an extended tip on the plug to allow semi-needle valve control of gases and liquids. Similar to CG-965 CHEM-VAC valve but with the arms bent to minimize hold-up of fluids. See CG-971 for replacement CHEM-CAP metering plug and CG-905 for glass portion only.