Learn More
Med Vet International Centesis Valve / Drain 16", J1339E, Each
Catalog No. 5000495618
Encompass_Preferred
Bivalve allows syringe to aspirate fluids i.e. chest drainage and then expel those fluids via another port so continually active centesis can be achieved without disconnecting. Suggest use J0282d1 Barbed Connector for chest drainage or feeding tube connection. Centesis Drainage Kit 16"(40cm) long
Catalog No. 50-004-95618 Supplier Med Vet International Supplier No. JOR1339E
Bivalve allows syringe to aspirate fluids i.e. chest drainage and then expel those fluids via another port so continually active centesis can be achieved without disconnecting. Suggest use J0282d1 Barbed Connector for chest drainage or feeding tube connection. Centesis Drainage Kit 16"(40cm) long