Learn More
Chemglass Life Sciences Valve, Chem-Vac, Hi-Vac, Low Hold Up, 0-4mm, Viton O-Ring
SDP
Catalog No. CG92901
Encompass_Preferred
Valve, Chem-Vac, Hi-Vac, Low Hold Up, 0-4mm, Viton O-Ring. Similar to CG-927 CHEM-VAC valve but with the arms bent to minimize hold up of fluids. Valve has TFE plug with three o-rings, the tip o-ring making the primary seal on the precision formed tapered seat. O-rings on the non-rotating shaft form an excellent dynamic seal between the TFE plug and glass barrel. See CG-935 for replacement plug, CG-537 for o-ring replacement kit, and CG-905 for glass portion only.
Catalog No. CG92901 Supplier Chemglass Life Sciences Supplier No. CG92901
Valve, Chem-Vac, Hi-Vac, Low Hold Up, 0-4mm, Viton O-Ring. Similar to CG-927 CHEM-VAC valve but with the arms bent to minimize hold up of fluids. Valve has TFE plug with three o-rings, the tip o-ring making the primary seal on the precision formed tapered seat. O-rings on the non-rotating shaft form an excellent dynamic seal between the TFE plug and glass barrel. See CG-935 for replacement plug, CG-537 for o-ring replacement kit, and CG-905 for glass portion only.