Learn More
Chemglass Life Sciences Secondary Valve Kit, All PTFE Ball Valve with 3/4" compression fittings, for use with Air Operated Drain Valves
SDP
Catalog No. 501531332
Encompass
Secondary Valve Kit, All PTFE Ball Valve with 3/4" compression fittings,for use with Air Operated Drain Valves. This kit will allow for emergency shut-off of the reactor contents in the unlikely event of valve failure.
Catalog No. 50-153-1332 Supplier Chemglass Life Sciences Supplier No. CG1968V10
Secondary Valve Kit, All PTFE Ball Valve with 3/4" compression fittings,for use with Air Operated Drain Valves. This kit will allow for emergency shut-off of the reactor contents in the unlikely event of valve failure.