Learn More
BURKLE INC Valve lance, V4A, length 21.65 in. (550 mm), Ø 1.26 in. (32 mm)
Catalog No. 5000026088
Encompass_Preferred
Sampler for all-layer samples of viscous, paste-like, semi-liquid substances, according to DIN 51750 pt. 3 and DIN EN ISO 15528. Different layers remain intact particularly with paste-like substances.
Catalog No. 50-000-26088 Supplier BURKLE INC Supplier No. 53591600
Sampler for all-layer samples of viscous, paste-like, semi-liquid substances, according to DIN 51750 pt. 3 and DIN EN ISO 15528. Different layers remain intact particularly with paste-like substances.