Learn More
Chemglass Life Sciences Valve, Mininert, 20-400 GPI
SDP
Catalog No. 50143867
Encompass_Preferred
Valve, Mininert, 20-400 GPI. These resealable valves allow repeated and unlimited syringe-needle access to samples. Slide the valve one way to open the needle-port. Slide the valve back to close and completely seal. Models are available for screw-cap and crimp-top vials. 20mm cap size.
Catalog No. 50-143-867 Supplier Chemglass Life Sciences Supplier No. CG4914020
Valve, Mininert, 20-400 GPI. These resealable valves allow repeated and unlimited syringe-needle access to samples. Slide the valve one way to open the needle-port. Slide the valve back to close and completely seal. Models are available for screw-cap and crimp-top vials. 20mm cap size.