Learn More
Trajan Scientific Valve MOVP Pneumatic Medical 1/EA
SDP
Catalog No. 5000494446
Encompass_Preferred
ON/OFF valve with pneumatic operation (MOVP) - L flow path with single outlet and 50 mm long stem
Catalog No. 50-004-94446 Supplier Trajan Scientific Supplier No. 1236091
ON/OFF valve with pneumatic operation (MOVP) - L flow path with single outlet and 50 mm long stem