Learn More
Trajan Scientific Valve MOVPT Pneumatic Medical 1/EA
SDP
Catalog No. 5000494136
Encompass_Preferred
ON/OFF valve with pneumatic operation (MOVPT) - T flow path with dual outlet and 100 mm short stem
Catalog No. 50-004-94136 Supplier Trajan Scientific Supplier No. 12361033
ON/OFF valve with pneumatic operation (MOVPT) - T flow path with dual outlet and 100 mm short stem