Learn More
AVANTOR MASTERFLEX (VWR LLC) Masterflex PTFE Multi-Turn Needle Valve, 1/8"" NPT(F), 316 SS Body, 2800 mL/min water
Catalog No. 501961054
Encompass
Maximum pressure is 75 psi; maximum temperature is 200¿>F (93¿>C). Wetted parts are body and O-ring made of PTFE and CTFE valve spindle. Choose aluminum (Al) or 316 stainless steel (SS) body. Orifice is 0.125".
Catalog No. 50-196-1054 Supplier AVANTOR MASTERFLEX (VWR LLC) Supplier No. MFLX0639382
Maximum pressure is 75 psi; maximum temperature is 200¿>F (93¿>C). Wetted parts are body and O-ring made of PTFE and CTFE valve spindle. Choose aluminum (Al) or 316 stainless steel (SS) body. Orifice is 0.125".