Learn More
Trajan Scientific VICI Valco Cheminert 10 Port Switching Valve, 0.25mm, PEEK (VAL.C2H-1340D)
Catalog No. 50635034
Encompass
VICI Valco VICI Valco Intruments PN- C2H-1340D, 5000 PSI, Valve Style- Cheminert, Ports- 10, Port Size- 0.25 mm, 2 Positions PEEK VICI Valco Replacement Components Stator PN- C-1C40H LPP Stator PN- 100.LCC2VS10PKS VICI Valco Rotor PN- C2-13R0H LPP Rotor PN-100.LCC2VR10PKS (NEW LPP PN: VAL.C2H-1340D). Made in the USA.
Catalog No. 50-635-034 Supplier Trajan Scientific Supplier No. PAL.VLV10PKSW
VICI Valco VICI Valco Intruments PN- C2H-1340D, 5000 PSI, Valve Style- Cheminert, Ports- 10, Port Size- 0.25 mm, 2 Positions PEEK VICI Valco Replacement Components Stator PN- C-1C40H LPP Stator PN- 100.LCC2VS10PKS VICI Valco Rotor PN- C2-13R0H LPP Rotor PN-100.LCC2VR10PKS (NEW LPP PN: VAL.C2H-1340D). Made in the USA.