Learn More
IDEX Health & Science Upchurch VALVE PEEK 4 WAY RGHT ANG FLOW
Catalog No. NC1920907
Encompass_Preferred
Idex Valves, Overall Length: 2.18 in., Swept Volume: 6.2 uL, Diameter Inner: 0.04 in., Overall Width: 0.66 in., Pressure: 500 psi
Catalog No. NC1920907 Supplier IDEX Health & Science Upchurch Supplier No. V101L
Idex Valves, Overall Length: 2.18 in., Swept Volume: 6.2 uL, Diameter Inner: 0.04 in., Overall Width: 0.66 in., Pressure: 500 psi