Learn More
Trajan Scientific VICI Valco Cheminert 6 Port Injection Valve, 0.40mm, PEEK (VAL.C2V-2346D-CTC)
Catalog No. 50635039
Encompass
VICI Valco VICI Valco Intruments PN- C2V-2346D-CTC, 5000 PSI, Valve Style- Cheminert, Ports- 6, Port Size- 0.40 mm, 2 Positions PEEK VICI Valco Replacement Components Stator PN- C2V-2C46 LPP Stator PN- 100.LCC2VSPK VICI Valco Rotor PN- C2-23R6 LPP Rotor PN-100.LCC2VRPK (NEW LPP PN: VAL.C2V-2346D-CTC). Thermo Part Number: 00950-01-00317. Made in the USA.
Catalog No. 50-635-039 Supplier Trajan Scientific Supplier No. PAL.VLV6PK
VICI Valco VICI Valco Intruments PN- C2V-2346D-CTC, 5000 PSI, Valve Style- Cheminert, Ports- 6, Port Size- 0.40 mm, 2 Positions PEEK VICI Valco Replacement Components Stator PN- C2V-2C46 LPP Stator PN- 100.LCC2VSPK VICI Valco Rotor PN- C2-23R6 LPP Rotor PN-100.LCC2VRPK (NEW LPP PN: VAL.C2V-2346D-CTC). Thermo Part Number: 00950-01-00317. Made in the USA.