Learn More
Trajan Scientific VICI Valco Cheminert 6 Port Injection Valve, MicroVolume, 0.25mm, PEEK (VAL.C2V-1346D-CTC)
Catalog No. 50635046
Encompass
VICI Valco VICI Valco Intruments PN- C2V-1346D-CTC, 5000 PSI, Valve Style- Cheminert, Ports- 6, Port Size- 0.25 mm, 2 Positions PEEK VICI Valco Replacement Components Stator PN- C2V-1C46 LPP Stator PN- 100.LCC2SNPK VICI Valco Rotor PN- C2- 13R6 LPP Rotor PN-100.LCC2VNRPK (NEW LPP PN: VAL.C2V-1346D-CTC). Thermo Part Number: 00950-01-00314. Made in the USA.
Catalog No. 50-635-046 Supplier Trajan Scientific Supplier No. PAL.VLVN6PK
VICI Valco VICI Valco Intruments PN- C2V-1346D-CTC, 5000 PSI, Valve Style- Cheminert, Ports- 6, Port Size- 0.25 mm, 2 Positions PEEK VICI Valco Replacement Components Stator PN- C2V-1C46 LPP Stator PN- 100.LCC2SNPK VICI Valco Rotor PN- C2- 13R6 LPP Rotor PN-100.LCC2VNRPK (NEW LPP PN: VAL.C2V-1346D-CTC). Thermo Part Number: 00950-01-00314. Made in the USA.