Learn More
Grainger ASCO Valve Repair Kit, Brass, Series 210, 1 ea
Catalog No. 502794814
Encompass_Preferred
This valve rebuild kit is designed for maintenance and restoration of 2-way, 2/2 normally closed general service solenoid valves. It includes essential components to replace worn internal parts, ensuring the continued performance and reliability of the valve assembly.
  • Includes bonnet, core assembly, core tube assembly, diaphragm, seat, and springs
  • Compatible with Series 210 models 8210B020 and 8210B021
  • Constructed with NBR sealing material
Catalog No. 50-279-4814 Supplier Grainger Supplier No. 1AKJ9
This valve rebuild kit is designed for maintenance and restoration of 2-way, 2/2 normally closed general service solenoid valves. It includes essential components to replace worn internal parts, ensuring the continued performance and reliability of the valve assembly.
  • Includes bonnet, core assembly, core tube assembly, diaphragm, seat, and springs
  • Compatible with Series 210 models 8210B020 and 8210B021
  • Constructed with NBR sealing material