Learn More
Grainger ASCO Valve Repair Kit, Solenoid Valve, 8210 Series Compatible, 1 ea
Catalog No. 502794819
Encompass_Preferred
This valve repair kit is designed for the maintenance and repair of compatible solenoid valves. The kit provides essential replacement components to restore valve function and ensure reliable operation in fluid control systems.
  • Includes cap, clip, core assembly, core tube assembly, diaphragm/seat assembly, O-rings, and springs
  • Compatible with 8210 series solenoid valves
  • UL listed
Catalog No. 50-279-4819 Supplier Grainger Supplier No. 1AKU6
This valve repair kit is designed for the maintenance and repair of compatible solenoid valves. The kit provides essential replacement components to restore valve function and ensure reliable operation in fluid control systems.
  • Includes cap, clip, core assembly, core tube assembly, diaphragm/seat assembly, O-rings, and springs
  • Compatible with 8210 series solenoid valves
  • UL listed