Learn More
Grainger ASCO Valve Repair Kit, For 8221G005 Solenoid Valve
Catalog No. 502794820
Encompass_Preferred
This solenoid valve rebuild kit provides essential replacement components for maintaining the performance and reliability of specified valves. The kit contains the parts subject to wear, helping to restore proper function and extend the service life of the valve assembly.
  • Includes cap, clip, core tube assembly, flow control, o-rings, piston assembly, seal, springs, support, and tube
  • Designed for 8221G005 normally closed valves
  • Includes detailed installation instructions
Catalog No. 50-279-4820 Supplier Grainger Supplier No. 1AKV1
This solenoid valve rebuild kit provides essential replacement components for maintaining the performance and reliability of specified valves. The kit contains the parts subject to wear, helping to restore proper function and extend the service life of the valve assembly.
  • Includes cap, clip, core tube assembly, flow control, o-rings, piston assembly, seal, springs, support, and tube
  • Designed for 8221G005 normally closed valves
  • Includes detailed installation instructions