Learn More
Chemglass Life Sciences 0-4mm Chem-Vac, Chem-Cap Valve Replacement Plug, Metering, Hi-Vac
SDP
Catalog No. CG97101
Encompass_Preferred
0-4mm Chem-Vac, Chem-Cap Valve Replacement Plug, Metering, Hi-Vac. Replacement CHEM-CAP metering plug and control knob for CHEM-VAC high vacuum valves CG-962 through CG-967. Plug is sold complete with control knob and CHEM-CAP metering stem. Control knob is not sold separately.
Catalog No. CG97101 Supplier Chemglass Life Sciences Supplier No. CG97101
0-4mm Chem-Vac, Chem-Cap Valve Replacement Plug, Metering, Hi-Vac. Replacement CHEM-CAP metering plug and control knob for CHEM-VAC high vacuum valves CG-962 through CG-967. Plug is sold complete with control knob and CHEM-CAP metering stem. Control knob is not sold separately.