Learn More
Andwin Scientific VALVE SAFETY CHECK ASSBLY EA
SDP
Catalog No. NC0514230
Encompass_Preferred
The spare safety check valve is for the pipetus pipet filler. The safety check valve performs as a first line of defense from media being accidentally drawn up too far. It is sold individually and separately from the pipet filler.
Catalog No. NC0514230 Supplier Andwin Scientific Supplier No. 9903202
Add to Cart
Add to Cart
The spare safety check valve is for the pipetus pipet filler. The safety check valve performs as a first line of defense from media being accidentally drawn up too far. It is sold individually and separately from the pipet filler.