Learn More
Trajan Scientific VICI Valco 4-Position Cheminert Selector Valve, 9 Port, 0.25mm (C5FH-1004D-CTC)
Catalog No. 50635083
Encompass
VICI Valco Valve Selection 4-Position, 5K PSI, 0.25mm Ports, Cheminert. [VICI Valco PN: C5FH-1004D-CTC] (NEW LPP PN: C5FH-1004D-CTC). Made in the USA.
Catalog No. 50-635-083 Supplier Trajan Scientific Supplier No. 100.SEL98
VICI Valco Valve Selection 4-Position, 5K PSI, 0.25mm Ports, Cheminert. [VICI Valco PN: C5FH-1004D-CTC] (NEW LPP PN: C5FH-1004D-CTC). Made in the USA.