Learn More
Thermo Scientific™ Valve with SMART Indicating Purifier (VIP)
Catalog No. 60180950
Change view
Click to view available options
Type:
Multi-turn Valve
Multi-turn Valve with Combi Filter
Quarter Turn Valve with Combi Filter
Quarter Turn Valve with Triple Filter
Replacement Combi Cartridge
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type
60-180-950 Multi-turn Valve
60-180-952 Quarter Turn Valve with Triple Filter
60-180-953 Quarter Turn Valve with Combi Filter
60-180-951 Multi-turn Valve with Combi Filter
60-180-941 Replacement Combi Cartridge
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 60-180-950 Supplier Thermo Scientific™ Supplier No. 60180950
Provide either flow control or positive shut off while maintaining gas purity for gasses 99.9999% pure or better
  • Provides a visual indication of contaminants in the gas stream while it is removing them, ensuring consistent performance of analytical processes
  • Safeguards the gas stream and process
  • Specially cleaned for all analytical applications and will not generate contaminates that consume the purifier
  • Patented Quick-Change Base to allow for easy change out of spent purifiers
  • Available in both a multiturn and quarter turn models
  • All regulators supplied with 1/8 in. stainless steel output adaptor
TRUSTED_SUSTAINABILITY
Specifications
Type Multi-turn Valve