Description
- Provides a visual indication of contaminants in the gas stream while it is removing them, ensuring consistent performance of analytical processes
- Safeguards the gas stream and process
- Specially cleaned for all analytical applications and will not generate contaminates that consume the purifier
- Patented Quick-Change Base to allow for easy change out of spent purifiers
- Available in both a multiturn and quarter turn models
- All regulators supplied with 1/8 in. stainless steel output adaptor
Specifications
Specifications
| Type | Quarter Turn Valve with Combi Filter |