Learn More
Waters Corp Waters Solenoid Valve, V3, Two-Way, HPLC System
Catalog No. 50818917 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This Solenoid Valve - V3 is a replacement two-way solenoid valve used in HPLC systems. It is utilized in the 2695 Separations Module and is similar to the V3 solenoid valve found in the 717 Autosampler.
  • Replacement V3 solenoid valve.
  • Two-way solenoid valve design.
  • Designed for HPLC systems.
  • Compatible with 2695 Separations Module.
  • Compatible with 2695 D Dissolution Separations Module.
  • Compatible with 717Plus Autosampler.
Catalog No. 50-818-917 Supplier Waters Corp Supplier No. 700001229
This Solenoid Valve - V3 is a replacement two-way solenoid valve used in HPLC systems. It is utilized in the 2695 Separations Module and is similar to the V3 solenoid valve found in the 717 Autosampler.
  • Replacement V3 solenoid valve.
  • Two-way solenoid valve design.
  • Designed for HPLC systems.
  • Compatible with 2695 Separations Module.
  • Compatible with 2695 D Dissolution Separations Module.
  • Compatible with 717Plus Autosampler.