Learn More
Grainger Vulcan Hart Valve, 90° Angle, 1/4 in. Mpt, 1/2 in.-27 Outlet, 1 ea
Catalog No. 502792764
Encompass_Preferred
This valve acts as a direct replacement component for ovens and warming equipment, designed to restore functionality to units requiring maintenance. It features a robust design suitable for specified cooking equipment models.
  • Angle configuration of 90°
  • Inlet connection is 1/4 in. MPT
  • Outlet connection measures 1/2 in.-27
  • Stem size of 0.240 in.
  • Overall height of 1 in.
  • Rotation functionality of off/on
  • Center inlet to end of stem distance of 2-1/8 in.
  • Stem position located on the right; stem flat is on the right when inlet MPT faces down
  • Compatible with range series Spr, Vsp 100, 200, 300, and WSPR 1, 2, 3 models
Catalog No. 50-279-2764 Supplier Grainger Supplier No. 19XL86
This valve acts as a direct replacement component for ovens and warming equipment, designed to restore functionality to units requiring maintenance. It features a robust design suitable for specified cooking equipment models.
  • Angle configuration of 90°
  • Inlet connection is 1/4 in. MPT
  • Outlet connection measures 1/2 in.-27
  • Stem size of 0.240 in.
  • Overall height of 1 in.
  • Rotation functionality of off/on
  • Center inlet to end of stem distance of 2-1/8 in.
  • Stem position located on the right; stem flat is on the right when inlet MPT faces down
  • Compatible with range series Spr, Vsp 100, 200, 300, and WSPR 1, 2, 3 models