Learn More
Trajan Scientific VALVE SS ON/OFF SMOV-1
SDP
Catalog No. 502530152
Encompass_Preferred
Trajan Stainless Steel Micro Control Valve, Valve Code: SMOV, Style: L Single Outlet manual, Connection Size: 0.06 in., 0.15 cm, Max. Pressure: 700 psi, Micro control valve, Temperature Maximum: 212 deg.F, Temperature Max: 100 deg.C
Catalog No. 50-253-0152 Supplier Trajan Scientific Supplier No. 1236283
Trajan Stainless Steel Micro Control Valve, Valve Code: SMOV, Style: L Single Outlet manual, Connection Size: 0.06 in., 0.15 cm, Max. Pressure: 700 psi, Micro control valve, Temperature Maximum: 212 deg.F, Temperature Max: 100 deg.C