Learn More
Chemglass Life Sciences Valve, Swabable, with 1/8" Barb
SDP
Catalog No. NC1923738
Encompass
The unique and innovative patented design allows the valve to be used in a wide variety of demanding applications where high flow and a closed system is a must. The valves are DEHP free, made with ISO-10993 materials, and not made with natural rubber latex. Bodies are made with BPA-free materials and polycarbonate. The valves mate securely with standard ISO-594 compliant syringes and connectors. Our barbed valve is Ideal for use as a sampling port in biopharmaceutical applications and are designed for easy assembly directly to tubing without the use of a Luer connector or solvents and adhesives.
Catalog No. NC1923738 Supplier Chemglass Life Sciences Supplier No. CLS139021V
The unique and innovative patented design allows the valve to be used in a wide variety of demanding applications where high flow and a closed system is a must. The valves are DEHP free, made with ISO-10993 materials, and not made with natural rubber latex. Bodies are made with BPA-free materials and polycarbonate. The valves mate securely with standard ISO-594 compliant syringes and connectors. Our barbed valve is Ideal for use as a sampling port in biopharmaceutical applications and are designed for easy assembly directly to tubing without the use of a Luer connector or solvents and adhesives.