Learn More
Hach Company Valve switching assembly - replacement switching valve for Lachat manifold (Hach 24927)
Catalog No. NC1952466 Shop All Hach Company Products
Encompass
Replacement switching valve assembly for manifold-based flow-injection systems. Designed for use with Lachat manifolds, this service part restores reliable flow switching in FIA analyzers and related instrumentation, helping maintain accurate sampling and reduce downtime.
  • Replacement part for manifold flow switching.
  • Restores valve function and sampling accuracy.
  • Compatible with flow-injection analysis manifolds.
  • Reduces instrument downtime during maintenance.
  • Durable construction for reliable operation.
Catalog No. NC1952466 Supplier Hach Company Supplier No. 24927
Replacement switching valve assembly for manifold-based flow-injection systems. Designed for use with Lachat manifolds, this service part restores reliable flow switching in FIA analyzers and related instrumentation, helping maintain accurate sampling and reduce downtime.
  • Replacement part for manifold flow switching.
  • Restores valve function and sampling accuracy.
  • Compatible with flow-injection analysis manifolds.
  • Reduces instrument downtime during maintenance.
  • Durable construction for reliable operation.