Learn More
Waters Corp Waters Switching Valve, Two-Position, Six-Port, Stainless Steel
Catalog No. 50793627 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This switching valve automates multiple injector and switching tasks, eliminating manual operation. It is designed for high throughput sample preparation and analysis, offering unattended operation and compatibility with various systems.
  • Eliminates manual switching valves.
  • Automates multiple injector and switching tasks.
  • Saves time with unattended operation.
  • Accelerates high throughput sample preparation and analysis.
  • Ready-to-use and compatible with any system.
  • Valve positions indicated by digital display or remote selection.
  • Built-in electronic communications for broad compatibility.
Catalog No. 50-793-627 Supplier Waters Corp Supplier No. WAT050046
This switching valve automates multiple injector and switching tasks, eliminating manual operation. It is designed for high throughput sample preparation and analysis, offering unattended operation and compatibility with various systems.
  • Eliminates manual switching valves.
  • Automates multiple injector and switching tasks.
  • Saves time with unattended operation.
  • Accelerates high throughput sample preparation and analysis.
  • Ready-to-use and compatible with any system.
  • Valve positions indicated by digital display or remote selection.
  • Built-in electronic communications for broad compatibility.