Learn More
IDEX Health & Science Upchurch VALVE TEFZ 2-WAY W/1/16 FITNG
Catalog No. NC1920884
Encompass_Preferred
Idex Shut-off Valves, Blue, Diameter Outer: 0.158 cm, 0.0625 in., Tefzel (ETFE), Pressure: 600 psig, Shut-Off Valve, Includes: 1/4-28 fittings for tubing
Catalog No. NC1920884 Supplier IDEX Health & Science Upchurch Supplier No. P782
Idex Shut-off Valves, Blue, Diameter Outer: 0.158 cm, 0.0625 in., Tefzel (ETFE), Pressure: 600 psig, Shut-Off Valve, Includes: 1/4-28 fittings for tubing