Learn More
Grainger Schneider Electric Hydronic Globe Valve, Bronze, 1/2 in., NPT x Union, 2.2 Cv
Catalog No. 502761086
Encompass_Preferred
The VB-7211 series is a two-way hydronic globe valve designed for proportional or two-position control of hot water, chilled water, or low-pressure saturated steam in HVAC systems. Featuring a durable bronze body with brass trim and a stainless steel stem, this valve provides reliable flow regulation for building climate control. It operates with a stem-up-open action and is engineered for precise performance in systems requiring equal percentage flow characteristics.
  • Flow coefficient (Cv) of 2.2
  • Temperature range of 20°F to 281°F (-7°C to 138°C)
  • Pressure rating up to 250 psi
  • 1/2 in. NPT x union end connections
  • Designed with a low-profile bronze body and spring-loaded TFE packing
  • Compatible with electric or pneumatic actuators
  • Not for use in combustible gas applications
Catalog No. 50-276-1086 Supplier Grainger Supplier No. 161Y76
The VB-7211 series is a two-way hydronic globe valve designed for proportional or two-position control of hot water, chilled water, or low-pressure saturated steam in HVAC systems. Featuring a durable bronze body with brass trim and a stainless steel stem, this valve provides reliable flow regulation for building climate control. It operates with a stem-up-open action and is engineered for precise performance in systems requiring equal percentage flow characteristics.
  • Flow coefficient (Cv) of 2.2
  • Temperature range of 20°F to 281°F (-7°C to 138°C)
  • Pressure rating up to 250 psi
  • 1/2 in. NPT x union end connections
  • Designed with a low-profile bronze body and spring-loaded TFE packing
  • Compatible with electric or pneumatic actuators
  • Not for use in combustible gas applications