Learn More
MSC CPC Colder Products MCD1304NA PTF Brass, Quick Disconnect, Valved Inline Coupling Body 250 Max psi, -40 to 180°F, 1.87" Overall Length x 0.63" Overall Height, Chrome Plated
Catalog No. NC1730608
Encompass
CPC Colder Products MCD1304NA PTF Brass, Quick Disconnect, Valved Inline Coupling Body 250 Max psi, -40 to 180°F, 1.87" Overall Length x 0.63" Overall Height, Chrome Plated
Catalog No. NC1730608 Supplier MSC Supplier No. 83051417
CPC Colder Products MCD1304NA PTF Brass, Quick Disconnect, Valved Inline Coupling Body 250 Max psi, -40 to 180°F, 1.87" Overall Length x 0.63" Overall Height, Chrome Plated