Learn More
Aviva Systems Biology Corp Vamp1 Antibody - C-terminal region (ARP59488_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP59488_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Vamp1. It was validated on Western Blot. . Alternate names: Syb1ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-286-717