Learn More
Aviva Systems Biology Corp Vamp7 Antibody - C-terminal region (ARP47593_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP47593_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Vamp7. It was validated on Western Blot. . Alternate names: Sybl1, TI-VAMP, VAMP-7ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Sheep, Zebrafish
Catalog No. 50-285-314