Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Supertek™ Van De Graff Generator
GREENER_CHOICESCI_ED

Catalog No. S139441 Shop All Supertek Scientific Products
Change view
Click to view available options
Grade:
1+
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Grade
S139441 1+
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S139441 Supplier Supertek™ Supplier No. PH45095
Add to Cart
Add to Cart

This Supertek™ Van De Graff Generator, is a small but powerful Van De Graaff generator. Generates high potential and sparks up to 3 in.

  • Aluminum 6 in. diameter dome.
  • Total height 18 in. with a ground terminal for discharge electrode.
  • Comes with one replacement belt.
  • Experiment book included.

Specifications

Grade 1+
Format Class Set
Languages English
Subject Physical Science - Electricity
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.