Learn More
Electron Microscopy Sciences Van Geisons Solution 250 ML
SDP
Catalog No. 5030079
Encompass_Preferred
Used in Modified Verhoff Elastic Van Gieson Stain for connective tissue stains – stain with a poly-acid step. See references: Gravey, Winsome et al "A modified Verhoff Elastic Van Gieson Stain" The Journal of Histotechnology, Vol. 14, No. 2, pp113 –114. June 1991.
Catalog No. 50-300-79 Supplier Electron Microscopy Sciences Supplier No. 2636905
Used in Modified Verhoff Elastic Van Gieson Stain for connective tissue stains – stain with a poly-acid step. See references: Gravey, Winsome et al "A modified Verhoff Elastic Van Gieson Stain" The Journal of Histotechnology, Vol. 14, No. 2, pp113 –114. June 1991.