Learn More
Electron Microscopy Sciences Van Gieson's Solution 500 ML
SDP
Supplier:  Electron Microscopy Sciences 2604605
Encompass_Preferred
Van Gieson's Solution is a prepared, ready-to-use, high quality staining solution that saves time and effort and produces consistent uniform results. Use with Weigert's Iodine or Weigert's Iron as a connective tissue stain.
Catalog No. 50-296-08